Learn More
Propargyl bromide, 97%, 80% w/w in toluene, stab. with magnesium oxide
Description
Propargyl bromide is used as an alkylating agent as well as an intermediate for the synthesis of organic compounds, agrochemicals and pharmaceuticals. It reacts with magnesium in presence of ether to get a Grignard reagent namely allenylmagnesium bromide, which on further reaction with propargyl bromide prepares bis-allene 1,2,4,5-hexatetraene. Also, used as metal brightening agents, corrosion inhibitor and soil fumigant.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Propargyl bromide is used as an alkylating agent as well as an intermediate for the synthesis of organic compounds, agrochemicals and pharmaceuticals. It reacts with magnesium in presence of ether to get a Grignard reagent namely allenylmagnesium bromide, which on further reaction with propargyl bromide prepares bis-allene 1,2,4,5-hexatetraene. Also, used as metal brightening agents, corrosion inhibitor and soil fumigant.
Solubility
Miscible with ethanol, ether, benzene, carbon tetrachloride and chloroform. Immiscible with water.
Notes
Light sensitive. Store in a cool place. Incompatible withbases, strong oxidizing agents, copper, peroxides, silver, silver oxides, mercury and mercury oxides.
Specifications
Specifications
| Melting Point | -61°C |
| Density | 1.335 |
| Boiling Point | 88°C to 90°C |
| Flash Point | 3°C (37°F) |
| Linear Formula | HC≡CCH2Br |
| Refractive Index | 1.494 |
| Quantity | 25 g |
| UN Number | UN2924 |
| Beilstein | 605309 |
| Sensitivity | Light sensitive |
| Show More |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.