missing translation for 'onlineSavingsMsg'
Learn More

Propargyl Bromide (Stablized with MgO), TRC

Product Code. 30426257
Click to view available options
Quantity:
50 g
Unit Size:
50g
This item is not returnable. View return policy

Product Code. 30426257

Brand: Toronto Research Chemicals TRCP76257550G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Propargyl Bromide
CAS 106-96-7
Molecular Formula C3H3Br
Quantity 50 g
Synonym 3-Bromopropyne, 1-Bromo-2-propyne, 2-Propynyl Bromide, 3-Bromo-1-propyne, 3-Bromopropyne, NSC 8801
InChI Formula InChI=1S/C3H3Br/c1-2-3-4/h1H,3H2
SMILES BrCC#C
IUPAC Name 3-bromoprop-1-yne
Molecular Weight (g/mol) 118.96
Formula Weight 117.9418
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.