missing translation for 'onlineSavingsMsg'
Learn More

Propargyl chloride, 98%

Product Code. 15274904 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15274904 5 g Glass bottle 5g
15264904 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15274904 Supplier Thermo Scientific Acros Supplier No. 451840050
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 624-65-7 | C3H3Cl | 74.51 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 624-65-7
Molecular Formula C3H3Cl
Molecular Weight (g/mol) 74.51
MDL Number MFCD00000980
InChI Key LJZPPWWHKPGCHS-UHFFFAOYSA-N
Synonym propargyl chloride, 3-chloropropyne, 3-chloro-1-propyne, 1-propyne, 3-chloro, 2-propynyl chloride, propyne, 3-chloro, propargylchloride, unii-3m62yfl252, gamma-chloroallylene, 1-chloro-2-propyne
PubChem CID 12221
IUPAC Name 3-chloroprop-1-yne
SMILES ClCC#C

Specifications

Density 1.0300g/mL
Boiling Point 58.0°C to 61.0°C
Flash Point 19°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula HC≡CCH2Cl
Quantity 5 g
Merck Index 15,7924
Specific Gravity 1.03
Solubility Information Solubility in water: insoluble.
Formula Weight 74.51
Percent Purity 98%
Chemical Name or Material Propargyl chloride
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Propargyl chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H301-Toxic if swallowed.
  • H330-Fatal if inhaled.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.