missing translation for 'onlineSavingsMsg'
Learn More

Propargylamine, 99%

Product Code. 10225330 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
Unit Size:
100g
1kg
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10225330 5 g 5g
10577141 25 g 25g
15224634 100 g 100g
15204634 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10225330 Supplier Thermo Scientific Acros Supplier No. 131460050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2450-71-7 | C3H5N | 55.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2450-71-7
Molecular Formula C3H5N
Molecular Weight (g/mol) 55.08
MDL Number MFCD00008198
InChI Key JKANAVGODYYCQF-UHFFFAOYSA-N
Synonym propargylamine, 2-propynylamine, 2-propyn-1-amine, 3-aminopropyne, 3-amino-1-propyne, 2-propyn-1-thiol, 3-aminoprop-1-yne, propargylamin, pro-pargylamine, propargyl amine
PubChem CID 239041
IUPAC Name prop-2-yn-1-amine
SMILES C#CCN

Specifications

Color Colorless to Yellow
Density 0.8600g/mL
Boiling Point 84.0°C
Flash Point 3°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula HCidentical toCCH2NH2
Refractive Index 1.4460 to 1.4500
Quantity 5 g
Beilstein 04,228
Specific Gravity 0.86
Solubility Information Solubility in water: miscible in water.
Formula Weight 55.08
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Propargylamine, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Propargylamine
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.