missing translation for 'onlineSavingsMsg'
Learn More

Propargylamine, TRC

Product Code. 30420412
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30420412 5 g
30426312 1 g
30416649 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30420412 Supplier Toronto Research Chemicals Supplier No. TRCP7625205G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2450-71-7
Molecular Formula C3 H5 N
Molecular Weight (g/mol) 55.08
Synonym 2-Propynylamine, 3-Aminoprop-1-yne, 3-Aminopropyne, N-(2-Propyn-1-yl)amine, NSC 80642
IUPAC Name prop-2-yn-1-amine
SMILES NCC#C

Specifications

Quantity 5 g
InChI Formula InChI=1S/C3H5N/c1-2-3-4/h1H,3-4H2
Formula Weight 55.0422
Chemical Name or Material Propargylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.