missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Propazine-d14 (di-iso-propyl-d14), CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Propazine D14 (Diisopropyl D14) |
| CAS | 1219802-87-5 |
| Molecular Formula | C9 D14 H2 Cl N5 |
| Quantity | 0.01 g |
| Synonym | Propazine D14 (Diisopropyl D14) |
| InChI Formula | InChI=1 S/C9H16ClN5/c1-5(2)11-8-13-7(10)14-9(15-8)12-6(3)4/h5-6 H,1-4H3,(H2,11,12,13,14,15)/i1D3,2D3,3D3,4D3,5 D,6 D |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])(Nc1nc(Cl)nc(NC([2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H])n1)C([2 H])([2 H])[2 H] |
| IUPAC Name | 6-chloro-2-N,4-N-bis(1,1,1,2,3,3,3-heptadeuteriopropan-2-yl)-1,3,5-triazine-2,4-diamine |
| Molecular Weight (g/mol) | 243.796 |
| Formula Weight | 243.197 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction