Learn More
Tocris Bioscience™ Propidium iodide
GREENER_CHOICE
Red-fluorescent DNA stain; membrane impermeant to live cells. Pi staining differentiates live and dead cells
Brand:  Tocris Bioscience™ 5135/10
Encompass
Product Code. 15796625
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Propidium iodide is a red-fluorescent DNA stain. Membrane impermeant to live cells. Propidium iodide staining differentiates between live and dead cells in cell populations.
Specifications
Specifications
95%
3,8-Diamino-5-[3-(diethylmethylammonio)propyl]-6-phenylphenanthridinium diiodide
-20°C
Fluorescent DNA Stains
25535-16-4