missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Propidium iodide

Product Code. 18760468
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18760468 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18760468 Supplier R&D Systems™ Supplier No. 5135/10
  • ONLINE EXCLUSIVE PRICE
    93.15 €
    84.00 € / 10mg
    Save 9.15 €
    9% Off
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Red-fluorescent DNA stain; membrane impermeant to live cells. Pi staining differentiates live and dead cells

Propidium iodide is a red-fluorescent DNA stain. Membrane impermeant to live cells. Propidium iodide staining differentiates between live and dead cells in cell populations.
TRUSTED_SUSTAINABILITY

Specifications

CAS 25535-16-4
Quantity 10 mg
Product Line Tocris
Molecular Formula 3,8-Diamino-5-[3-(diethylmethylammonio)propyl]-6-phenylphenanthridinium diiodide
Product Type Propidium iodide
Purity 95%
Molecular Weight (g/mol) M.W. (Observed): 668.39
Inhibitors Propidium iodide

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.