Learn More
STEMCELL Technologies Propidium Iodide
Product Code. 17180352
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Product Code. Quantity Unit Size
17180352 10 mg 10mg
1 options
Product Code. 17180352 Supplier STEMCELL Technologies Supplier No. 75002
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Cell viability dye (DNA-labeling dye)
  • Propidium Iodide (PI) is a red-fluorescent cell viability dye which is excluded from live cells with intact membranes, but penetrates dead or damaged cells and binds to DNA and RNA by intercalating between the bases
  • It is widely used as a counterstain to differentiate and exclude non-viable cells in flow cytometric analyses, and can be excited using blue (488 nm), green (532 nm), or yellow-green (561 nm) laser lines, with detection in the FL2 or FL3 channels
  • PI is used in DNA fluorescence imaging applications to discriminate early and late stages of apoptosis, to study cell‐mediated cytotoxicity, and for chromosome analysis
  • It is also commonly used in quantitative DNA assays
Specifications
CAS 25535-16-4
Quantity 10 mg