missing translation for 'onlineSavingsMsg'
Learn More

Propiolic acid, 98+%

Product Code. 11470783
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11470783 100 g 100g
11450783 5 g 5g
11460783 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11470783 Supplier Thermo Scientific Alfa Aesar Supplier No. A13245.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 471-25-0 | C3H2O2 | 70.05 g/mol

Propiolic acid is a reagent used in the preparation of transition metal complexes, haloalkyl propiolates and halopropenoates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Propiolic acid is a reagent used in the preparation of transition metal complexes, haloalkyl propiolates and halopropenoates.

Solubility
Miscible with water, ether, ethanol and chloroform.

Notes
Store in cool place. Incompatible with bases, oxidizing agents and reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 471-25-0
Molecular Formula C3H2O2
Molecular Weight (g/mol) 70.05
MDL Number MFCD00004360
InChI Key UORVCLMRJXCDCP-UHFFFAOYSA-N
Synonym propiolic acid, 2-propynoic acid, propynoic acid, propargylic acid, acetylenecarboxylic acid, carboxyacetylene, propiolic acidd, propiolicacid, unii-p2qw39g9lz, propynoate
PubChem CID 10110
ChEBI CHEBI:33199
IUPAC Name prop-2-ynoic acid
SMILES OC(=O)C#C

Specifications

Melting Point 18°C
Density 1.136
Boiling Point 102°C (200 mmHg)
Flash Point 58°C (136°F)
Refractive Index 1.433
Quantity 100 g
UN Number UN2920
Beilstein 878176
Merck Index 14,7824
Solubility Information Miscible with water,ether,ethanol and chloroform.
Formula Weight 70.05
Percent Purity ≥98%
Chemical Name or Material Propiolic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.