missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Propionaldehyde-2,2-d2, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.10g
0.25g
Specifications
Specifications
| Chemical Name or Material | Propionaldehyde-2,2-d2 |
| CAS | 39493-21-5 |
| Molecular Formula | C3 D2 H4 O |
| Quantity | 0.25 g |
| Synonym | Propanal-2,2-d2 (9 CI), 2,2-Dideuteropropanal |
| InChI Formula | InChI=1 S/C3H6O/c1-2-3-4/h3H,2H2,1H3/i2D2 |
| SMILES | [2 H]C([2 H])(C)C=O |
| IUPAC Name | 2,2-dideuteriopropanal |
| Molecular Weight (g/mol) | 60.09 |
| Formula Weight | 60.0544 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction