missing translation for 'onlineSavingsMsg'
Learn More

Propionaldehyde (>80%), TRC

Product Code. 30427124
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30427124 5 g 5g
30410875 10 g -
30415466 25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30427124 Supplier Toronto Research Chemicals Supplier No. TRCP7825155G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-38-6
Molecular Formula C3 H6 O
Molecular Weight (g/mol) 58.08
Synonym Propanal, Propionaldehyde (7CI,8CI), 1-Propanal, Ethylcarboxaldehyde, Methylacetaldehyde, NSC 6493, Propaldehyde, Propanaldehyde, Propional, Propionic aldehyde, Propylaldehyde, Propylic aldehyde, n-Propanal, n-Propionaldehyde
IUPAC Name propanal
SMILES CCC=O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C3H6O/c1-2-3-4/h3H,2H2,1H3
Formula Weight 58.0419
Chemical Name or Material Propionaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.