missing translation for 'onlineSavingsMsg'
Learn More

Propionaldehyde, 97%

Product Code. 11413207
Change view
Click to view available options
Quantity:
500 mL
1000 mL
5000 mL
Unit Size:
1000mL
5000mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11413207 5000 mL 5000mL
11474010 500 mL 500mL
11403207 1000 mL 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 11413207 Supplier Thermo Scientific Alfa Aesar Supplier No. A16146.0J
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 123-38-6 | C3H6O | 58.08 g/mol

Propionaldehyde applications include as an intermediate, in pharmaceuticals, perfumes, plastics and polyols or polyhydric alcohols. It is also used as a precursor to trimethylolethane (CH3C(CH2OH)3) through a condensation reaction with formaldehyde, this triol is an important intermediate in the production of alkyd resins. Other applications include reduction to propanol and oxidation to propionic acid. Also used in PEGylated protein purification.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Propionaldehyde applications include as an intermediate, in pharmaceuticals, perfumes, plastics and polyols or polyhydric alcohols. It is also used as a precursor to trimethylolethane (CH3C(CH2OH)3) through a condensation reaction with formaldehyde, this triol is an important intermediate in the production of alkyd resins. Other applications include reduction to propanol and oxidation to propionic acid. Also used in PEGylated protein purification.

Solubility
Fully miscible in water.

Notes
Air sensitive. Incompatible with oxidizing agents and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-38-6
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00007020
InChI Key NBBJYMSMWIIQGU-UHFFFAOYSA-N
Synonym propionaldehyde, propanaldehyde, propaldehyde, propional, propionic aldehyde, n-propanal, methylacetaldehyde, propylaldehyde, propylic aldehyde, aldehyde propionique
PubChem CID 527
ChEBI CHEBI:17153
IUPAC Name propanal
SMILES CCC=O

Specifications

Melting Point -81°C
Density 0.81
Boiling Point 47°C to 49°C
Flash Point −40°C (−40°F)
Odor Pungent
Refractive Index 1.362
Quantity 5000 mL
UN Number UN1275
Beilstein 506010
Sensitivity Air sensitive
Merck Index 14,7823
Solubility Information Fully miscible in water.
Formula Weight 58.08
Percent Purity 97%
Chemical Name or Material Propionaldehyde
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.