missing translation for 'onlineSavingsMsg'
Learn More

Propionaldehyde, TRC

Product Code. 30007898
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30007898 10 g 10g
30007703 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30007898 Supplier Toronto Research Chemicals Supplier No. TRCP78250010G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-38-6
Molecular Formula C3 H6 O
Molecular Weight (g/mol) 58.08
Synonym Propanal, Propionaldehyde (7CI,8CI), 1-Propanal, Ethylcarboxaldehyde, Methylacetaldehyde, NSC 6493, Propaldehyde, Propanaldehyde, Propional, Propionic aldehyde, Propylaldehyde, Propylic aldehyde, n-Propanal, n-Propionaldehyde
IUPAC Name propanal
SMILES CCC=O

Specifications

Quantity 10 g
InChI Formula InChI=1S/C3H6O/c1-2-3-4/h3H,2H2,1H3
Formula Weight 58.0419
Chemical Name or Material Propionaldehyde
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.