missing translation for 'onlineSavingsMsg'
Learn More

Propionamidoxime, 97%

Product Code. 11449590
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11449590 1 g 1g
11439590 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11449590 Supplier Thermo Scientific Alfa Aesar Supplier No. H50889.03
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 29335-36-2 | C3H8N2O | 88.11 g/mol

Propionamidoxime is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Propionamidoxime is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
It is hygroscopic. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29335-36-2
Molecular Formula C3H8N2O
Molecular Weight (g/mol) 88.11
MDL Number MFCD03426272
InChI Key RLZPCFQNZGINRP-UHFFFAOYSA-N
Synonym n-hydroxypropionamidine, n-hydroxypropionimidamide, 1z-n'-hydroxypropanimidamide, 1-hydroxyimino propylamine, n-hydroxy-propionamidine, n-hydroxyproponmdamde, n-hydroxypropanamidine, n'-hydroxypropimidamide, z-n'-hydroxypropimidamide, e-n'-hydroxypropanimidamide
PubChem CID 6521662
IUPAC Name N'-hydroxypropanimidamide
SMILES CCC(=NO)N

Specifications

Quantity 1 g
Sensitivity Hygroscopic
Solubility Information Slightly soluble in water.
Formula Weight 88.11
Percent Purity 97%
Chemical Name or Material Propionamidoxime

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.