missing translation for 'onlineSavingsMsg'
Learn More

Propionic acid, 99%, pure

Product Code. p-4510137 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 mL
1 L
2.5 L
10 L
Packaging:
EcoSafPak™ Glass Bottle
Glass Bottle
Plastic drum
Unit Size:
1 L
10 L
2.5 L
5mL
This item is not returnable. View return policy

Product Code. 10386690

Brand: Thermo Scientific Acros 149300050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 79-09-4 | C3H6O2 | 74.079 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-09-4
InChI Key XBDQKXXYIPTUBI-UHFFFAOYSA-N
Synonym propionic acid, ethylformic acid, methylacetic acid, carboxyethane, ethanecarboxylic acid, pseudoacetic acid, metacetonic acid, monoprop, luprosil, prozoin
PubChem CID 1032
ChEBI CHEBI:30768
IUPAC Name propanoic acid
SMILES CCC(=O)O

Specifications

Density 0.9900g/mL
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Molecular Formula C3H6O2
Linear Formula C2H5CO2H
Refractive Index 1.3850 to 1.3870 (20°C, 589nm)
Quantity 5 mL
Beilstein 02, 234
Fieser 12,415
Merck Index 15, 7939
Molecular Weight (g/mol) 74.079
Formula Weight 74.08
Vapor Pressure 5mbar at 20°C
Percent Purity ≥98.5% (GC)
Grade Pure
Physical Form Liquid
Chemical Name or Material Propionic acid
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.