missing translation for 'onlineSavingsMsg'
Learn More

Propionic acid, 99%

Product Code. 11356846
Change view
Click to view available options
Quantity:
100 g
1000 g
2500 g
Unit Size:
100g
1kg
2.5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11356846 2500 g 2.5kg
11336846 100 g 100g
11346846 1000 g 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11356846 Supplier Thermo Scientific Alfa Aesar Supplier No. L04210.0E
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 79-09-4 | C3H6O2 | 74.079 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Propionic acid is used as a preservative for animal feed and food for human consumption. It acts as an intermediate in the production of polymers like cellulose-acetate-propionate, artificial flavorings, agrochemicals and in bleaching agents. In addition, it is used in the synthesis of morphans.

Solubility
Miscible with water, alcohol, diethyl ether and chloroform.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-09-4
Molecular Formula C3H6O2
Molecular Weight (g/mol) 74.079
MDL Number MFCD00002756
InChI Key XBDQKXXYIPTUBI-UHFFFAOYSA-N
Synonym propionic acid, ethylformic acid, methylacetic acid, carboxyethane, ethanecarboxylic acid, pseudoacetic acid, metacetonic acid, monoprop, luprosil, prozoin
PubChem CID 1032
ChEBI CHEBI:30768
IUPAC Name propanoic acid
SMILES CCC(=O)O

Specifications

Melting Point -21°C
Density 0.992
Boiling Point 140°C to 141°C
Flash Point 51°C (123°F)
Odor Unpleasant
Linear Formula CH3CH2CO2H
Refractive Index 1.3875
Quantity 2500 g
UN Number UN3463
Beilstein 506071
Merck Index 14,7825
Solubility Information Miscible with water,alcohol,diethyl ether and chloroform.
Formula Weight 74.08
Percent Purity 99%
Chemical Name or Material Propionic acid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Propionic acid
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.