missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifikationer
Specifikationer
| CAS | 79-09-4 |
| Melting Point | -22°C |
| pH | 2.5 |
| Boiling Point | 141°C |
| Packaging | HDPE plastic bottle |
| Molecular Formula | C3H6O2 |
| MDL Number | 2756 |
| Quantity | 500 mL |
| Synonym | propionic acid, ethylformic acid, methylacetic acid, carboxyethane, ethanecarboxylic acid, pseudoacetic acid, metacetonic acid, monoprop, luprosil, prozoin |
| InChI Key | XBDQKXXYIPTUBI-UHFFFAOYSA-N |
| Visa mer |
Säkerhet och hantering
Produktbeteckning
- Propionic acid (low UV absorption)
- Danger
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
- H226-Flammable liquid and vapour.
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contain: propanoic acid
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?