missing translation for 'onlineSavingsMsg'
Learn More

Propionic acid, sodium salt, 99.0-100.5%

Product Code. 10081550 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
1 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
Unit Size:
1kg
25g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10081550 25 g Plastic bottle 25g
10245340 1 kg Plastic bottle 1kg
10423965 5 kg Plastic drum 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10081550 Supplier Thermo Scientific Acros Supplier No. 149010250
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 137-40-6 | C3H5NaO2 | 96.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 137-40-6
Molecular Formula C3H5NaO2
Molecular Weight (g/mol) 96.06
MDL Number MFCD00002759
InChI Key JXKPEJDQGNYQSM-UHFFFAOYSA-M
Synonym sodium propionate, sodium propanoate, propionic acid sodium salt, propanoic acid, sodium salt, napropion, ocuseptine, deketon, impedex, keenate, propiofar
PubChem CID 2723816
IUPAC Name sodium;propanoate
SMILES CCC(=O)[O-].[Na+]

Specifications

Melting Point 285°C to 286°C
Color White
Flash Point >250°C
Infrared Spectrum Authentic
Assay Percent Range 99.0-100.5%
Packaging Plastic bottle
Linear Formula C2H5CO2Na
Quantity 25 g
Beilstein 02, 234
Merck Index 15, 8798
Solubility Information Solubility in water: very soluble. Other solubilities: soluble in alcohol
Formula Weight 96.06
Percent Purity 99.0-100.5%
Physical Form Crystalline Powder
Chemical Name or Material Propionic acid, sodium salt
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.