missing translation for 'onlineSavingsMsg'
Learn More

Propionic Anhydride-d10, CDN

Product Code. 30327214
Change view
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30327214 1 g 1g
30324467 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30327214 Supplier CDN Supplier No. CDND01571G

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Propionic Anhydride-d10
CAS 870471-31-1
Molecular Formula C6 D10 O3
Quantity 1 g
Synonym 1,1'-Anhydride Propanoic Acid-d10, Anhydride Propanoic Acid-d10, Methylacetic Anhydride-d10, Propanoic Anhydride-d10, Propionic Acid Anhydride-d10, Propanoic-d5 acid, anhydride (9 CI), Propanoic-d5 acid, anhydride (9 CI), Propanoic-d5 acid, anhydride (9 CI), Propanoic-d5 acid, anhydride (9 CI), Propanoic-d5 acid, anhydride (9 CI), Propanoic-d5 acid, anhydride (9 CI), Propanoic-d5 acid, anhydride (9 CI)
InChI Formula InChI=1 S/C6H10O3/c1-3-5(7)9-6(8)4-2/h3-4H2,1-2H3/i1D3,2D3,3D2,4D2
SMILES [2 H]C([2 H])([2 H])C([2 H])([2 H])C(=O)OC(=O)C([2 H])([2 H])C([2 H])([2 H])[2 H]
IUPAC Name 2,2,3,3,3-pentadeuteriopropanoyl 2,2,3,3,3-pentadeuteriopropanoate
Molecular Weight (g/mol) 140.2
Formula Weight 140.1258 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.