missing translation for 'onlineSavingsMsg'
Learn More

Propionic Anhydride-d10, TRC

Product Code. 30412741
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30412741 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30412741 Supplier Toronto Research Chemicals Supplier No. TRCP786382100MG

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Propionic Anhydride-d10
CAS 870471-31-1
Molecular Formula C6 D10 O3
Quantity 100 mg
Synonym 1,1'-Anhydride Propanoic Acid-d10, Anhydride Propanoic Acid-d10, Methylacetic Anhydride-d10, Propanoic Anhydride-d10, Propionic Acid Anhydride-d10, Propionyl Anhydride-d10, Propionyl Oxide-d10, Propanoic-2,2,3,3,3-d5 acid','1,1'-anhydride, Propanoic-d5 acid','anhydride (9CI), D10-Propionyl anhydride, Propionic anhydride D10, Propanoyl-d5 propanoate-d5
InChI Formula InChI=1S/C6H10O3/c1-3-5(7)9-6(8)4-2/h3-4H2,1-2H3/i1D3,2D3,3D2,4D2
SMILES [2H]C([2H])([2H])C([2H])([2H])C(=O)OC(=O)C([2H])([2H])C([2H])([2H])[2H]
IUPAC Name 2,2,3,3,3-pentadeuteriopropanoyl 2,2,3,3,3-pentadeuteriopropanoate
Molecular Weight (g/mol) 140.2
Formula Weight 140.1258
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.