missing translation for 'onlineSavingsMsg'
Learn More

Propionitrile, 99%

Product Code. 11424634
Change view
Click to view available options
Quantity:
250 mL
1000 mL
Unit Size:
1000mL
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11424634 250 mL 250mL
11414634 1000 mL 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11424634 Supplier Thermo Scientific Alfa Aesar Supplier No. A13203.AK
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 107-12-0 | C3H5N | 55.08 g/mol

Propionitrile, act as solvents and are reacted further for various application including, Extraction solvent for fatty acids, oils and unsaturated hydrocarbons, Solvent for spinning and casting, removing agent of colouring matters and aromatic alcohols, Non-aqueous solvent for titrations and for inorganic salts, Recrystallization of steroids, Parent compound for organic synthesis, chemical intermediate in biochemistry, Catalyst and component of transition-metal complex catalysts, Stabilizer for chlorinated solvents, Chemical intermediate and solvent for perfumes and pharmaceuticals. It is used as a dielectric fluid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Propionitrile, act as solvents and are reacted further for various application including, Extraction solvent for fatty acids, oils and unsaturated hydrocarbons, Solvent for spinning and casting, removing agent of colouring matters and aromatic alcohols, Non-aqueous solvent for titrations and for inorganic salts, Recrystallization of steroids, Parent compound for organic synthesis, chemical intermediate in biochemistry, Catalyst and component of transition-metal complex catalysts, Stabilizer for chlorinated solvents, Chemical intermediate and solvent for perfumes and pharmaceuticals. It is used as a dielectric fluid.

Solubility
It decomposes with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.Take measures to prevent the build up of electrostatic charge. Incompatible with Strong acids, Strong bases, Strong oxidizing agents, Strong reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-12-0
Molecular Formula C3H5N
Molecular Weight (g/mol) 55.08
MDL Number MFCD00001948
InChI Key FVSKHRXBFJPNKK-UHFFFAOYSA-N
Synonym propionitrile, ethyl cyanide, propiononitrile, cyanoethane, propylnitrile, propionic nitrile, ether cyanatus, hydrocyanic ether, ethylkyanid, propannitril
PubChem CID 7854
ChEBI CHEBI:26307
IUPAC Name propanenitrile
SMILES CCC#N

Specifications

Melting Point -92°C
Density 0.781
Boiling Point 97°C to 98°C
Flash Point 6°C (42°F)
Odor Pleasant
Refractive Index 1.366
Quantity 250 mL
UN Number UN2404
Beilstein 773680
Merck Index 14,7827
Solubility Information It decomposes with water.
Formula Weight 55.08
Percent Purity 99%
Chemical Name or Material Propionitrile
Show More Show Less
compliance-icons compliance-icons
Produktidentifikator
  • Propionitrile
Signalwort
  • Danger
Gefahrenkategorie
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
Gefahrenhinweis
  • H225-Highly flammable liquid and vapour.
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H319-Causes serious eye irritation.
  • H331-Toxic if inhaled.
Sicherheitshinweis
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
Ergänzende Informationen
  • MIXTURE LIST-Contain: Propionitrile

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.