missing translation for 'onlineSavingsMsg'
Learn More

Propionyl bromide, 95%

Product Code. 10146373 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10146373 5 g Glass bottle 5g
10413125 25 g Glass bottle 25g
10024293 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10146373 Supplier Thermo Scientific Acros Supplier No. 392360050
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 598-22-1 | C3H5BrO | 136.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 598-22-1
Molecular Formula C3H5BrO
Molecular Weight (g/mol) 136.98
MDL Number MFCD00000116
InChI Key RIBFXMJCUYXJDZ-UHFFFAOYSA-N
Synonym propionyl bromide, propanoyl-bromide, propionylbromide, propanyl bromide, acmc-209mg3, ksc270q9p, ribfxmjcuyxjdz-uhfffaoysa, inchi=1/c3h5bro/c1-2-3 4 5/h2h2,1h3
PubChem CID 69018
IUPAC Name propanoyl bromide
SMILES CCC(=O)Br

Specifications

Color Blue-Gray
Density 1.5200g/mL
Boiling Point 104°C
Flash Point 52°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH2COBr
Refractive Index 1.4525 to 1.4575 (20°C, 589nm)
Quantity 5 g
Specific Gravity 1.52
Solubility Information Solubility in water: reacts violently
Formula Weight 136.98
Percent Purity 95%
Physical Form Tablets
Chemical Name or Material Propionyl bromide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Propionyl bromide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H226-Flammable liquid and vapour.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.