missing translation for 'onlineSavingsMsg'
Learn More

Propionyl chloride, 99.5%, phosgene free

Product Code. 15565996 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
500 g
Unit Size:
25g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15565996 500 g 500g
15555996 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 15565996 Supplier Thermo Scientific Acros Supplier No. 458975000
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 79-03-8 | C3H5ClO | 92.52 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-03-8
Molecular Formula C3H5ClO
Molecular Weight (g/mol) 92.52
InChI Key RZWZRACFZGVKFM-UHFFFAOYSA-N
Synonym propionyl chloride, propionic acid chloride, propionic chloride, propionylchloride, unii-mb6vl5omb9, mb6vl5omb9, propanoylchloride, propionyl cloride, propanoic chloride, propionyl-chloride
PubChem CID 62324
IUPAC Name propanoyl chloride
SMILES CCC(=O)Cl

Specifications

Melting Point -94.0°C
Density 1.0590g/mL
Boiling Point 80.0°C
Flash Point 6°C
Infrared Spectrum Authentic
Assay Percent Range 99.4% min. (GC)
Linear Formula C2H5COCl
Quantity 500 g
Specific Gravity 1.059
Solubility Information Solubility in water: reacts.
Viscosity 0.48 mPa.s (20°C)
Formula Weight 92.52
Percent Purity 99.5%
Chemical Name or Material Propionyl chloride
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Propionyl chloride, phosgene free
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.