missing translation for 'onlineSavingsMsg'
Learn More

Propiophenone, 99%

Product Code. 10051470 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2 mL
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
2.5 L
250mL
2mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10051470 1 L Glass bottle 1 L
10715791 2 mL Glass bottle 2mL
10439060 250 mL Glass bottle 250mL
11934851 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 10051470 Supplier Thermo Scientific Acros Supplier No. 131540010
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 93-55-0 | C9H10O | 134.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93-55-0
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.18
InChI Key KRIOVPPHQSLHCZ-UHFFFAOYSA-N
Synonym propiophenone, ethyl phenyl ketone, propionylbenzene, 1-phenyl-1-propanone, 1-propanone, 1-phenyl, phenyl ethyl ketone, ketone, ethyl phenyl, propionphenone, usaf ek-1235, 1-phenyl-propan-1-one
PubChem CID 7148
ChEBI CHEBI:425902
IUPAC Name 1-phenylpropan-1-one
SMILES CCC(=O)C1=CC=CC=C1

Specifications

Melting Point 18.0°C
Color Colorless to Yellow
Density 1.0000g/mL
Boiling Point 218.0°C
Flash Point 87°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5COC2H5
Refractive Index 1.5248 to 1.5268
Quantity 1 L
Beilstein 07,II,231
Merck Index 15,7944
Specific Gravity 1
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with methanol, ether, benzene and toluene, insoluble in glycerol, ethylene glycol and, propylene glycol
Formula Weight 134.18
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Propiophenone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.