missing translation for 'onlineSavingsMsg'
Learn More

Propyl Beta-D-Glucuronide, TRC

Product Code. 30011182
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011182 5 mg 5mg
30137863 10 mg 10mg
30004303 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011182 Supplier Toronto Research Chemicals Supplier No. TRCP8352755MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17685-07-3
Molecular Formula C9 H16 O7
Molecular Weight (g/mol) 236.22
Synonym Propyl β-D-Glucopyranosiduronic Acid;
IUPAC Name (2S,3S,4S,5R,6R)-3,4,5-trihydroxy-6-propoxyoxane-2-carboxylic acid
SMILES CCCO[C@@H]1O[C@@H]([C@@H](O)[C@H](O)[C@H]1O)C(=O)O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C9H16O7/c1-2-3-15-9-6(12)4(10)5(11)7(16-9)8(13)14/h4-7,9-12H,2-3H2,1H3,(H,13,14)/t4-,5-,6+,7-,9+/m0/s1
Formula Weight 236.0896
Chemical Name or Material Propyl beta-D-Glucuronide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.