missing translation for 'onlineSavingsMsg'
Learn More

Propyl gallate, 98%

Product Code. 10659843 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10659843 2.5 kg Plastic bottle 2.5kg
10459250 100 g Plastic bottle 100g
10625762 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10659843 Supplier Thermo Scientific Acros Supplier No. 131580025
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 121-79-9 | C10H12O5 | 212.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-79-9
Molecular Formula C10H12O5
Molecular Weight (g/mol) 212.2
MDL Number MFCD00002196
InChI Key ZTHYODDOHIVTJV-UHFFFAOYSA-N
Synonym propyl gallate, n-propyl gallate, progallin p, tenox pg, nipagallin p, gallic acid, propyl ester, gallic acid propyl ester, nipa 49, benzoic acid, 3,4,5-trihydroxy-, propyl ester, 3,4,5-trihydroxybenzene-1-propylcarboxylate
PubChem CID 4947
ChEBI CHEBI:10607
IUPAC Name propyl 3,4,5-trihydroxybenzoate
SMILES CCCOC(=O)C1=CC(=C(C(=C1)O)O)O

Specifications

Melting Point 146.0°C to 150.0°C
Color White to Beige
Loss on Drying 0.5% max. (110°C, 4 hrs)
Infrared Spectrum Authentic
Packaging Plastic bottle
Quantity 2.5 kg
Merck Index 15, 7971
Solubility Information Solubility in water: 0.35g/100 mL (25°C). Other solubilities: 103g/100 g alcohol,83g/100 g ether,soluble in aceton,propyleenglycol and,polysorbaat 8
Formula Weight 212.2
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Propyl gallate, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Propyl gallate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.