missing translation for 'onlineSavingsMsg'
Learn More

Propyl Gallate, TRC

Product Code. 30416700
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30416700 10 g
30410254 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30416700 Supplier Toronto Research Chemicals Supplier No. TRCP83700010G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-79-9
Molecular Formula C10 H12 O5
Molecular Weight (g/mol) 212.2
Synonym Propyl Gallate, 3,4,5-Trihydroxybenzoic acid propyl ester, Gallic acid propyl ester, Antioxidant PG, E 310, Gallic acid n-propyl ester, Hemostyptin, Marupi Gallate, NSC 2626, Nipa 49, Nipagallin P, Nipanox S 1, PG, Progallin P, Propyl 3,4,5-trihydroxybenzoate, Propyl gallate, Tenox PG, n-Propyl 3,4,5-trihydroxybenzoate, n-Propyl gallate
IUPAC Name propyl 3,4,5-trihydroxybenzoate
SMILES CCCOC(=O)c1cc(O)c(O)c(O)c1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C10H12O5/c1-2-3-15-10(14)6-4-7(11)9(13)8(12)5-6/h4-5,11-13H,2-3H2,1H3
Formula Weight 212.0685
Percent Purity >95%
Chemical Name or Material Gallic acid-propyl ester
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.