missing translation for 'onlineSavingsMsg'
Learn More

Propylamine, 99+%, extra pure

Product Code. 10052920 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 mL
500 mL
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
2.5 L
500mL
50mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10052920 1 L Glass bottle 1 L
10723031 50 mL Glass bottle 50mL
10697192 500 mL Glass bottle 500mL
13418478 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10052920 Supplier Thermo Scientific Acros Supplier No. 220610010
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 107-10-8 | C3H9N | 59.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-10-8
Molecular Formula C3H9N
Molecular Weight (g/mol) 59.11
MDL Number MFCD00008205
InChI Key WGYKZJWCGVVSQN-UHFFFAOYSA-N
Synonym propylamine, 1-propanamine, 1-propylamine, propanamine, n-propylamine, 1-aminopropane, monopropylamine, mono-n-propylamine, propyl amine, n-propyl amine
PubChem CID 7852
ChEBI CHEBI:39870
IUPAC Name propan-1-amine
SMILES CCCN

Specifications

Melting Point -83.0°C
Density 0.7190g/mL
Boiling Point 48.0°C
Flash Point −30°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH2CH2NH2
Refractive Index 1.3870 to 1.3890
Quantity 1 L
Beilstein 04,136
Fieser 13,164
Merck Index 15,7956
Specific Gravity 0.719
Solubility Information Solubility in water: soluble in water. Other solubilities: miscible with alcohol and ether
Viscosity 0.4 mPa.s (20°C)
Formula Weight 59.11
Percent Purity 99+%
Grade Extra Pure
Physical Form Liquid
Chemical Name or Material Propylamine, 99+%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Propylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H290-May be corrosive to metals.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.