missing translation for 'onlineSavingsMsg'
Learn More

Propylamine hydrochloride, 98%

Product Code. 10511091 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10511091 25 g Glass bottle 25g
10042850 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10511091 Supplier Thermo Scientific Acros Supplier No. 221890250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 556-53-6 | C3H10ClN | 95.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 556-53-6
Molecular Formula C3H10ClN
Molecular Weight (g/mol) 95.57
MDL Number MFCD00012911
InChI Key PYNUOAIJIQGACY-UHFFFAOYSA-N
Synonym propylamine hydrochloride, n-propylamine hydrochloride, 1-propanamine, hydrochloride, propylammonium chloride, propan-1-amine hydrochloride, 1-propylammonium chloride, n-propylammonium chloride, propylamine, hydrochloride, unii-a1z6ovy2hg, a1z6ovy2hg
PubChem CID 11165
IUPAC Name propan-1-amine;hydrochloride
SMILES [H+].[Cl-].CCCN

Specifications

Melting Point 160°C to 164°C
Color Beige to White
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula CH3CH2CH2NH2·HCl
Quantity 25 g
Beilstein 04, 137
Merck Index 15, 7956
Formula Weight 95.57
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material Propylamine hydrochloride, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • Propylamine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.