missing translation for 'onlineSavingsMsg'
Learn More

Propylene carbonate, 99.5%, anhydrous, AcroSeal™

Product Code. 13177714 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
100mL
800mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
13177714 800 mL AcroSeal™ Glass Bottle 800mL
13118663 100 mL AcroSeal™ Glass Bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 13177714 Supplier Thermo Scientific Acros Supplier No. 429268000
  • ONLINE EXCLUSIVE PRICE
    120.00 €
    116.00 € / 800mL
    Save 4.00 €
    3% Off
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 108-32-7 | C4H6O3 | 102.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-32-7
Molecular Formula C4H6O3
Molecular Weight (g/mol) 102.09
MDL Number MFCD00005385,MFCD00798264,MFCD00798265
InChI Key RUOJZAUFBMNUDX-UHFFFAOYNA-N
Synonym propylene carbonate, 1,2-propylene carbonate, 1,2-propanediol cyclic carbonate, texacar pc, 1,3-dioxolan-2-one, 4-methyl, cyclic propylene carbonate, arconate 5000, 1,2-propanediol carbonate, 1-methylethylene carbonate, cyclic 1,2-propylene carbonate
PubChem CID 7924
IUPAC Name 4-methyl-1,3-dioxolan-2-one
SMILES CC1COC(=O)O1

Specifications

Melting Point -49.0°C
Density 1.2040g/mL
Boiling Point 240.0°C to 243.0°C
Flash Point 116°C
Assay Percent Range 99.4% min. (GC)
Packaging AcroSeal™ Glass Bottle
Refractive Index 1.4200 to 1.4220
Quantity 800 mL
Specific Gravity 1.204
Solubility Information Solubility in water: 200g/L (25°C). Other solubilities: miscible with acetone,benzene,chloroform,dmf,,ether,ethyl acetate and ethyl alcohol,slightly soluble in carbon tetrachloride,soluble in most organic solvents
Viscosity 2.8 mPa.s (20°C)
Formula Weight 102.09
Percent Purity 99.5%
Chemical Name or Material Propylene carbonate
Show More Show Less
compliance-icons
Product Identifier
  • Propylene carbonate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.