missing translation for 'onlineSavingsMsg'
Learn More

Propylene Carbonate, 99%, Reagent Grade, Honeywell™

Product Code. 15688030 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
2.5 L
This item is not returnable. View return policy

Product Code. 15688030

Brand: Honeywell Research Chemicals P526521L

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Reagent Grade, 99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-32-7
Molecular Formula C4H6O3
Molecular Weight (g/mol) 102.09
MDL Number MFCD00005385,MFCD00798264,MFCD00798265
InChI Key RUOJZAUFBMNUDX-UHFFFAOYNA-N
Synonym propylene carbonate, 1,2-propylene carbonate, 1,2-propanediol cyclic carbonate, texacar pc, 1,3-dioxolan-2-one, 4-methyl, cyclic propylene carbonate, arconate 5000, 1,2-propanediol carbonate, 1-methylethylene carbonate, cyclic 1,2-propylene carbonate
PubChem CID 7924
IUPAC Name 4-methyl-1,3-dioxolan-2-one
SMILES CC1COC(=O)O1

Specifications

Melting Point -55°C
Density 1.204g/mL at 25°C
pH 7 (at 200g/L 20°C)
Boiling Point 240°C
Ignition Point 851°F
Odor Fruit like
Packaging Glass bottle
Quantity 1 L
UN Number NONH for all modes of transport
Beilstein 107913
Formula Weight 102.09g/mol
Vapor Pressure 0.13mmHg (20°C), 0.98mmHg (50°C)
Percent Purity 0.99
Chemical Name or Material Propylene carbonate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.