missing translation for 'onlineSavingsMsg'
Learn More

Propylene Carbonate, TRC

Product Code. 30138026
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138026 1 g 1g
30134128 50 g 50g
17920685 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138026 Supplier Toronto Research Chemicals Supplier No. TRCP8367401G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-32-7
Molecular Formula C4 H6 O3
Molecular Weight (g/mol) 102.09
Synonym 1,3-Dioxolan-2-one, 4-methyl-, Carbonic acid, cyclic propylene ester (8CI), Carbonic acid, propylene ester (6CI), 4-Methyl-1,3-dioxolan-2-one, 1,2-Propanediol carbonate, 1,2-Propanediol cyclic carbonate, 1,2-Propanediyl carbonate, 1,2-Propylene carbonate, 1-Methylethylene carbonate, 2-Methyl-1,2-ethylene carbonate, 2-Oxo-4-methyl-1,3-dioxolane, 4-Methyl-1,3-dioxolane-2-one, 4-Methyl-2-dioxolanone, 4-Methyl-2-oxo-1,3-dioxolane, Arconate 1000, Arconate 5000, Arconate HP, Arconate propylenecarbonate, Carbonic acid cyclic 1,2-propylene ester, Carbonic acid cyclic methylethylene ester, Cyclic 1,2-propylene carbonate, Cyclic methylethylene carbonate, Cyclic propylene carbonate, Fluor Solvent, JN 02, Jeffsol 1555, Jeffsol AG 1555, Jeffsol PC, Methyl-1,3-dioxolan-2-one, NSC 11784, NSC 1913, PC Medion, PC-HP, Propylene carbonate, Propylene glycol cyclic carbonate, Texacar PC
IUPAC Name 4-methyl-1,3-dioxolan-2-one
SMILES CC1COC(=O)O1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C4H6O3/c1-3-2-6-4(5)7-3/h3H,2H2,1H3
Formula Weight 102.0317
Chemical Name or Material 1,2-Propylene carbonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.