missing translation for 'onlineSavingsMsg'
Learn More

Propylene Glycol, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Product Code. 15518834 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Narrow Mouth Amber Glass Bottle
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15518834 500 mL Narrow Mouth Amber Glass Bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15518834 Supplier Avantor J.T.Baker Supplier No. U51007
  • ONLINE EXCLUSIVE PRICE
    419.96 €
    342.33 € / 500mL
    Save 77.63 €
    18% Off
 Restricted Item
Estimated Shipment: 23-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 57-55-6
Molecular Formula C3H8O2
Molecular Weight (g/mol) 76.095
InChI Key DNIAPMSPPWPWGF-UHFFFAOYSA-N
Synonym 1,2-propanediol, propylene glycol, 1,2-dihydroxypropane, 2-hydroxypropanol, 1,2-propylene glycol, methylethyl glycol, methylethylene glycol, monopropylene glycol, isopropylene glycol, dowfrost
PubChem CID 1030
ChEBI CHEBI:16997
IUPAC Name propane-1,2-diol
SMILES CC(CO)O

Specifications

Quantity 500 mL
Formula Weight 76.1
Flash Point 99.00°C
Grade BAKER ANALYZED™ ACS Reagent
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 1.04kg
Chemical Name or Material Propylene Glycol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.