missing translation for 'onlineSavingsMsg'
Learn More

Propylene Glycol, Multi-Compendial, U.S.P., J.T. Baker™

Product Code. 15518614 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
4 L
19 L
Packaging:
Narrow Mouth Amber Glass Bottle
Poly Pail
Unit Size:
19 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15518614 19 L Poly Pail 19 L
15508614 4 L Narrow Mouth Amber Glass Bottle 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15518614 Supplier Avantor J.T.Baker Supplier No. 940307
 Restricted Item
Estimated Shipment: 02-03-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 57-55-6
Molecular Formula C3H8O2
Molecular Weight (g/mol) 76.095
InChI Key DNIAPMSPPWPWGF-UHFFFAOYSA-N
Synonym 1,2-propanediol, propylene glycol, 1,2-dihydroxypropane, 2-hydroxypropanol, 1,2-propylene glycol, methylethyl glycol, methylethylene glycol, monopropylene glycol, isopropylene glycol, dowfrost
PubChem CID 1030
ChEBI CHEBI:16997
IUPAC Name propane-1,2-diol
SMILES CC(CO)O

Specifications

Quantity 19 L
Flash Point 99.00°C
Packaging Poly Pail
Specific Gravity 1L = 1.04kg
Formula Weight 76.1
Percent Purity ≥99.5%
Grade Multi-Compendial/USP
Chemical Name or Material Propylene Glycol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.