missing translation for 'onlineSavingsMsg'
Learn More

(±)-Propylene oxide, 99.5%, extra pure, AcroSeal™

Product Code. 10084287 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
100mL
800mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10084287 800 mL AcroSeal™ Glass Bottle 800mL
10572138 100 mL AcroSeal™ Glass Bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10084287 Supplier Thermo Scientific Acros Supplier No. 430868000
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 75-56-9 | C3H6O | 58.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-56-9
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00005126
InChI Key GOOHAUXETOMSMM-UHFFFAOYNA-N
Synonym propylene oxide, 1,2-epoxypropane, methyloxirane, epoxypropane, propyleneoxide, propene oxide, oxirane, methyl, 1,2-propylene oxide, propylene epoxide, 2,3-epoxypropane
PubChem CID 6378
ChEBI CHEBI:38685
IUPAC Name 2-methyloxirane
SMILES CC1CO1

Specifications

CAS Min % 99.4
CAS Max % 100.0
Melting Point -112.0°C
Density 0.8300g/mL
Boiling Point 34.0°C
Flash Point −37°C
Infrared Spectrum Authentic
Assay Percent Range 99.4% min. (GC)
Packaging AcroSeal™ Glass Bottle
Refractive Index 1.3650 to 1.3670
Quantity 800 mL
Merck Index 15,7969
Specific Gravity 0.83
Solubility Information Solubility in water: 40g/100mL (20°C). Other solubilities: miscible with alcohol and ether
Viscosity 0.32 mPa.s (20°C)
Formula Weight 58.08
Percent Purity 99.5%
Grade Extra Pure
Physical Form Liquid
Chemical Name or Material (±)-Propylene oxide, Extra Pure
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.