missing translation for 'onlineSavingsMsg'
Learn More

(±)-Propylene oxide, >99%

Product Code. 11367348
Change view
Click to view available options
Quantity:
100 mL
1 L
3 x 1 L
Unit Size:
1 L
100mL
3 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11367348 100 mL 100mL
11387348 1 L 1 L
11377348 3 x 1 L 3 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11367348 Supplier Thermo Scientific Alfa Aesar Supplier No. 030765.AE
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 75-56-9 | C3H6O | 58.08 g/mol

Propylene oxide is used as a precursor in the production of polyether polyols, which is utilized for making polyurethane plastics, polypropylene glycols, propylene glycol ethers and propylene carbonate. It is involved in the preparation of biological samples for electron microscopy. It plays a vital role in the removal of residual ethanol previously used for dehydration. Further, it is used in thermobaric weapons, and microbial fumigation. It is also used as solvents, and in the preparation of surfactants, lubricants and in oil demulsifiers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-56-9
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00005126
InChI Key GOOHAUXETOMSMM-UHFFFAOYNA-N
Synonym propylene oxide, 1,2-epoxypropane, methyloxirane, epoxypropane, propyleneoxide, propene oxide, oxirane, methyl, 1,2-propylene oxide, propylene epoxide, 2,3-epoxypropane
PubChem CID 6378
ChEBI CHEBI:38685
IUPAC Name 2-methyloxirane
SMILES CC1CO1

Specifications

Melting Point °C to 112°C
Density 0.83 g/mL
Boiling Point 34°C to 35°C
Flash Point -37°C (-35°F)
Assay Percent Range >99%
Refractive Index 1.366
Quantity 100 mL
UN Number UN1280
Beilstein 79763
Merck Index 14,7856
Solubility Information Miscible with water,acetone,benzene,carbon tetrachloride,methanol and ether.
Formula Weight 58.08
Percent Purity ≥99%
Physical Form Liquid
Chemical Name or Material Propylene oxide
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • (+/-)-Propylene oxide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 1
  • Germ cell mutagenicity Category 1B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H224-Extremely flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H319-Causes serious eye irritation.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H340-May cause genetic defects.
  • H350-May cause cancer.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P311-Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contain: Propylene oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.