missing translation for 'onlineSavingsMsg'
Learn More

(±)-Propylene oxide, >99%

Product Code. 11377348
Click to view available options
Quantity:
100 mL
1 L
3 x 1 L
Unit Size:
1 L
100mL
3 L
This item is not returnable. View return policy

Product Code. 11377348

Brand: Thermo Scientific Alfa Aesar 030765.L7

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 75-56-9 | C3H6O | 58.08 g/mol

Propylene oxide is used as a precursor in the production of polyether polyols, which is utilized for making polyurethane plastics, polypropylene glycols, propylene glycol ethers and propylene carbonate. It is involved in the preparation of biological samples for electron microscopy. It plays a vital role in the removal of residual ethanol previously used for dehydration. Further, it is used in thermobaric weapons, and microbial fumigation. It is also used as solvents, and in the preparation of surfactants, lubricants and in oil demulsifiers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 75-56-9
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00005126
InChI Key GOOHAUXETOMSMM-UHFFFAOYNA-N
Synonym propylene oxide, 1,2-epoxypropane, methyloxirane, epoxypropane, propyleneoxide, propene oxide, oxirane, methyl, 1,2-propylene oxide, propylene epoxide, 2,3-epoxypropane
PubChem CID 6378
ChEBI CHEBI:38685
IUPAC Name 2-methyloxirane
SMILES CC1CO1

Specifikationer

Melting Point -112°C
Color Colorless
Density 0.83 g/mL
Boiling Point 34°C to 35°C
Flash Point -37°C (-35°F)
Assay Percent Range >99%
Odor Ether-like
Refractive Index 1.366
Quantity 3 x 1 L
UN Number UN1280
Beilstein 79763
Merck Index 14,7856
Solubility Information Miscible with water,acetone,benzene,carbon tetrachloride,methanol and ether.
Formula Weight 58.08
Percent Purity ≥99%
Physical Form Liquid
Chemical Name or Material (+/-)-Propylene oxide
Visa mer Visa mindre
compliance-icons compliance-icons compliance-icons
Produktbeteckning
  • (+/-)-Propylene oxide
Signalord
  • Danger
Riskkategorier
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 1
  • Germ cell mutagenicity Category 1B
  • Specific target organ toxicity Category 3
Faroangivelser
  • H224-Extremely flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H319-Causes serious eye irritation.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H340-May cause genetic defects.
  • H350-May cause cancer.
Skyddsangivelse
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P311-Call a POISON CENTER/doctor/ .
Kompletterande information
  • MIXTURE LIST-Contains : Propylene oxide

RUO – Research Use Only

Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.