missing translation for 'onlineSavingsMsg'
Learn More

(±)-Propylene Oxide, TRC

Product Code. 30412211
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30412211 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30412211 Supplier Toronto Research Chemicals Supplier No. TRCP835241500ML

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Propylene oxide
CAS 75-56-9
Molecular Formula C3 H6 O
Quantity 500 mL
Synonym Propylene Oxide, Oxirane','2-methyl-, Oxirane','methyl- (9CI), Oxypropylene (6CI), Propane','1,2-epoxy- (7CI), Propylene oxide (8CI), 2-Methyloxirane, (±)-1,2-Epoxypropane, (±)-2-Methyloxirane, (±)-Epoxypropane, (±)-Methyloxirane, (±)-Propylene oxide, 1,2-Epoxypropane, 1,2-Propylene oxide, 2,3-Epoxypropane, AD 6, AD 6 (suspending agent), Chemogas NV, DL-1,2-Epoxypropane, Epihydrin, Epoxypropane, Methyloxacyclopropane, Methyloxirane, Propene oxide, Propozone, Propylene epoxide, dl-Propylene oxide, rac-Propylene oxide
InChI Formula InChI=1S/C3H6O/c1-3-2-4-3/h3H,2H2,1H3
SMILES CC1CO1
IUPAC Name 2-methyloxirane
Molecular Weight (g/mol) 58.08
Formula Weight 58.0419
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.