missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ProQuantum™ Immunoassays Accessory Kit

Product Code. 15960493
Click to view available options
Quantity:
10 assays
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15960493

Brand: Invitrogen™ A36723

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The ProQuantum Immunoassay Accessory Kit provides the common consumable plastics to be used with any ProQuantum immunoassay product, conveniently bundled for first-time users who may be uncertain of what is needed or low-volume users who do not wish to buy these plastics in bulk.

Sufficient materials are provided to run 10 assays using 96-well plates. The kit does not include the following items: qPCR plates, cold blocks, and pipette tips.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Immunoassays Accessory Kit
Quantity 10 assays
Product Line ProQuantum
Content And Storage
  • 1 96-well plate rack
  • 10 25-mL reagent reservoirs for low volume pipetting
  • 20 adhesive seals
  • 10 optical seals
  • 1 hand seal applicator tool
  • 10 1.5-mL microfuge tubes
  • 10 15-mL conical tubes
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.