Learn More
DuPont™ ProShield™ 60
Product Code. 16254511 Shop All DuPont Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
Unit Size:
Pack of 50
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
16254511 Large Pack of 50
16234511 Small Pack of 50
16244511 Medium Pack of 50
16264511 X-Large Pack of 50
16274511 2X-Large Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 16254511 Supplier DuPont™ Supplier No. D15519554
  • ONLINE EXCLUSIVE PRICE
    397.66 €
    262.14 € / Pack of 50
    Save 135.52 €
    34% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
ProShield™ 60 provides a barrier against non-hazardous dry particles down to one micron in size.
  • Constructed of a microporous film on a polypropylene nonwoven fabric.
  • Combine good particle protection with high liquid repellency.
  • Help provide a barrier against non-hazardous dry particles down to one micron in size.
  • The material is permeable to moisture vapour only and is not air permeable.
  • Certified according to Regulation (EU) 2016/425.
  • Chemical protective clothing, Category III, Type 5 and 6.
  • EN 1073-2 (protection against radioactive contamination).
  • Antistatic treatment (EN 1149-5) on inside.
  • Stitched internal seams.
  • Nylon zipper with flap.
  • Ideal choice for applications that are less demanding in terms of barrier, durability and comfort.
TRUSTED_SUSTAINABILITY
Specifications
Size Large
Gender Unisex
Material Polypropylene
Color White
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Closure Type Zipper with Storm Flap
Seam Type Stitched
Sterility Non-sterile
Description Individually Packed
Quantity 50/Cs.