missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ ProShield™ 20 White

Product Code. 15411255 Shop All DuPont Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 50
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15411255 3X-Large Pack of 50
15461245 Small Pack of 50
15471245 Medium Pack of 50
15481245 Large Pack of 50
15491245 X-Large Pack of 50
15401255 2X-Large Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 15411255 Supplier DuPont™ Supplier No. D15338160
  • ONLINE EXCLUSIVE PRICE
    317.51 €
    222.89 € / Pack of 50
    Save 94.62 €
    29% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Designed to protect workers from certain substances with a high level of comfort. Dupont™ ProShield™ 20 White Hooded Coverall is typically used for protection against particles (Type 5) and limited liquid splashes or sprays (Type 6).

  • Chemical protective clothing, Category III, Type 5 and 6
  • EN 1073-2 (protection against radioactive contamination)
  • Antistatic treatment (EN 1149-5) - on inside
  • Stitched external seams
  • Nylon zipper with flap
  • Available in white in sizes S to XXXL (a blue garment is also available).
  • 2-piece hood.
  • Elasticated face, wrists, waist and ankles
TRUSTED_SUSTAINABILITY

Specifications

Product Type Hooded Coveralls
Size 3X-Large
Material SMS Fabric
Color White
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Closure Type Zipper with Storm Flap
Seam Type Stitched
Protection Class Category III, Type 5 and 6
Certifications/Compliance (EU) 2016/425, EN 1073-2, EN 1149-5
Size (Metric) Chest 124 to 132 cm
For Use With (Application) Workers seeking protection against dirt and grime during light duty work and other industries.
Quantity 50/Pk.
Show More Show Less

The user should make the risk assessment to determine the protection required.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.