missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Prostratin, 99+%

Product Code. 15403649
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
Product Code. Quantity unitSize
15403649 5 mg 5mg
15413649 1 mg 1mg
2 options
This item is not returnable. View return policy

Product Code. 15403649

Brand: Thermo Scientific Chemicals J60971.#0

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

12-Deoxyphorbol 13-acetate; activator of protein kinase C. A weak mouse ear irritant, but lethal at 1ug per mouse

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60857-08-1
Molecular Formula C22H30O6

Specifications

Quantity 5 mg
Description White
Content And Storage -30°C to -10°C
For Use With (Application) 12-Deoxyphorbol 13-acetate; activator of protein kinase C. A weak mouse ear irritant, but lethal at 1μg per mouse
Formula Weight 390.47
Melting Point 216°C to 219°C
Solubility Soluble in DMSO at 30mg/ml
Form Powder
InChI Key BOJKFRKNLSCGHY-KKFGZBDXSA-N
SMILES C[C@@H]1CC2(OC(C)=O)C([C@@H]3C=C(CO)C[C@@]4(O)[C@@H](C=C(C)C4=O)[C@@]13O)C2(C)C
IUPAC Name (1R,2S,6R,10S,15R)-1,6-dihydroxy-8-(hydroxymethyl)-4,12,12,15-tetramethyl-5-oxotetracyclo[8.5.0.0²,⁶.0¹¹,¹³]pentadeca-3,8-dien-13-yl acetate
Molecular Weight (g/mol) 390.48
Assay 99+%
Show More Show Less

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.