Learn More
Trajan Scientific and Medical™ ProteCol™ PEEK Fingertight Fittings
Product Code. 15219223
Change view
Click to view available options
Thread Size:
1/32 in. to 6-40 UNF
Unit Size:
Pack of 5
Product Code. Thread Size unitSize
15219223 1/32 in. to 6-40 UNF Pack of 5
1 options
Product Code. 15219223 Supplier Trajan Scientific and Medical™ Supplier No. 222017
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
Precision engineered for capillary LC. Trajan Scientific and Medical ProteCol™ PEEK Fingertight Fittings offer complete versatility for connecting capillary tubing.
  • Leak free connections
  • Replacement parts such as nuts and ferrules
Specifications
Type PEEK Fingertight Fittings 1/32 in. to 6-40UNF
Thread Size 1/32 in. to 6-40 UNF
Material Stainless Steel