missing translation for 'onlineSavingsMsg'
Learn More

3M™ Protective Coverall 4530

Product Code. 17815676
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
Unit Size:
Pack of 20
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
17815676 Medium Pack of 20
17835676 X-Large Pack of 20
17825676 2X-Large Pack of 20
11993774 Large Pack of 20
17844175 Small Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 17815676 Supplier 3M™ Supplier No. 7000109011
  • ONLINE EXCLUSIVE PRICE
    263.00 €
    174.38 € / Pack of 20
    Save 88.62 €
    33% Off
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Protective Coverall 4530 has a specialist material treatment which offers fabric flame spread resistance to allow the garment to be worn over an Index 2/3 FR garment, enhancing the chemical splash and hazardous dust protection and extending the life cycle through reduced laundering.

  • Use 3M™ Protective Coverall 4530 for protection against dirt, splashes and toxic dusts, including nuclear particles.
  • It meets a variety of standards for chemical protective clothing including EN 1149 Antistatic protection, EN 1073-2 Nuclear particulate resistance, EN ISO 13688, Type 6 Liquid Splash EN 13034 and Type 5 Dust EN ISO 13982-1.
  • You can identify it easily with the white triangle in the blue suit which is made of the same 54 gsm SMMS as the rest.
  • This coverall includes a specialist treatment of the material only (not components) which offers flame spread resistance complying with ISO 14116 Index 1.
  • This means the disposable coverall can be used with an FR Index 2/3 garment to offer improved certain chemical splash and hazardous dust performance and extend the life of the reusable garment by minimizing the need to launder.
  • The 3M™ protective coverall 4530 is soft and breathable yet robust and strong weighing in at 54 gsm.
  • It's made with SMMS which is a layer of spunbond on the inner and outer and 2 layers of meltblown material in the middle.
  • The Spunbond acts as the smooth strength and splash protection, while the Meltblown layers provide an intricate multilayered web of microfibres that filter out dry particulates.
  • Soft, breathable and very lightweight, the non-woven material allows for easier release of body heat and moisture, giving extended comfort wear for increased productivity in the workplace.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Protective Coveralls
Size Medium
Gender Unisex
Material SMS Polypropylene
Color Blue
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Knit
Neck Style Hooded
Hood Type 3-panel
Closure Type 2-way Zipper with Storm Flap
Seam Type Serged
Sterility Non-sterile
Protection Class Category III
Resistance Flame, Chemicals, Liquid, Anti-static
Certifications/Compliance Nuclear Particulate Resistance (EN 1073-2), Type 5 Dust (EN ISO 13982-1), Type 6 Liquid Splash (EN 13034), Flame-Spread Resistant (ISO 14116 Index 1), and Antistatic (EN 1149)
No. of Pockets None
Width (English) Chest 36.2 to 39.4 in.
Width (Metric) Chest 92 to 100 cm
Height (English) 65.7 to 69.3 in.
Height (Metric) 167 to 176 cm
Weight (Metric) 54 gsm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.