Learn More
3M™ Protective Coverall 4530
Product Code. 17844175 Shop All 3M Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
Unit Size:
Pack of 20
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
17844175 Small Pack of 20
17815676 Medium Pack of 20
17835676 X-Large Pack of 20
17825676 2X-Large Pack of 20
11993774 Large Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 17844175 Supplier 3M™ Supplier No. 7100214328
  • ONLINE EXCLUSIVE PRICE
    263.00 €
    203.52 € / Pack of 20
    Save 59.48 €
    22% Off
Estimated Shipment: 11-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Protective Coverall 4530 has a specialist material treatment which offers fabric flame spread resistance to allow the garment to be worn over an Index 2/3 FR garment, enhancing the chemical splash and hazardous dust protection and extending the life cycle through reduced laundering.
  • Use 3M™ Protective Coverall 4530 for protection against dirt, splashes and toxic dusts, including nuclear particles.
  • It meets a variety of standards for chemical protective clothing including EN 1149 Antistatic protection, EN 1073-2 Nuclear particulate resistance, EN ISO 13688, Type 6 Liquid Splash EN 13034 and Type 5 Dust EN ISO 13982-1.
  • You can identify it easily with the white triangle in the blue suit which is made of the same 54 gsm SMMS as the rest.
  • This coverall includes a specialist treatment of the material only (not components) which offers flame spread resistance complying with ISO 14116 Index 1.
  • This means the disposable coverall can be used with an FR Index 2/3 garment to offer improved certain chemical splash and hazardous dust performance and extend the life of the reusable garment by minimizing the need to launder.
  • The 3M™ protective coverall 4530 is soft and breathable yet robust and strong weighing in at 54 gsm.
  • It's made with SMMS which is a layer of spunbond on the inner and outer and 2 layers of meltblown material in the middle.
  • The Spunbond acts as the smooth strength and splash protection, while the Meltblown layers provide an intricate multilayered web of microfibres that filter out dry particulates.
  • Soft, breathable and very lightweight, the non-woven material allows for easier release of body heat and moisture, giving extended comfort wear for increased productivity in the workplace.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Protective Coveralls
Size Small
Gender Unisex
Material SMS Polypropylene
Color Blue
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Knit
Neck Style Hooded
Hood Type 3-panel
Closure Type 2-way Zipper with Storm Flap
Seam Type Serged
Sterility Non-sterile
Protection Class Category III
Resistance Flame, Chemicals, Liquid, Anti-static
Certifications/Compliance Nuclear Particulate Resistance (EN 1073-2), Type 5 Dust (EN ISO 13982-1), Type 6 Liquid Splash (EN 13034), Flame-Spread Resistant (ISO 14116 Index 1), and Antistatic (EN 1149)
No. of Pockets None