missing translation for 'onlineSavingsMsg'
Learn More

3M™ Protective Coverall 4565

Product Code. 17845666 Shop All 3M Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Unit Size:
Pack of 20
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
17845666 Medium Pack of 20
17855676 Small Pack of 20
17805666 Large Pack of 20
17865666 X-Large Pack of 20
17875666 2X-Large Pack of 20
17845676 3X-Large Pack of 20
17875676 4X-Large Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 17845666 Supplier 3M™ Supplier No. 7100256062
  • ONLINE EXCLUSIVE PRICE
    431.00 €
    283.41 € / Pack of 20
    Save 147.59 €
    34% Off
View all promotions
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Protective Coverall 4565 has desirable comfort features and provides a strong barrier to certain chemicals, liquid sprays and particles.

  • Wear 3M™ Protective Coverall 4565 for an ideal barrier to hazardous particles, limited liquid splashes and sprays to Type 4
  • Fresh white coverall 4565 has recognizable red taped seams and meets a variety of standards, including chemical protective clothing EN 14126 Infective agents, EN 1073-2 Nuclear particulate resistance, EN 1149, EN ISO 13688, Type 6 Liquid Splash EN 13034 and Type 5 Dust EN ISO 13982-1, and Type 4 Liquid Spray EN 14605
  • Taped seams reduce skin exposure points through tearing and reduced risk of inward leakage by chemicals or particles
  • Offers Til class 2 in Particulate Radioactive Contamination EN1073-2, meaning it has very good particle hold out as well as liquid spray resistance
  • Made with a non-microporous PE laminate for chemical protection with permeation-tested resistance against certain chemicals
  • Knitted cuffs, elasticated waist and ankles make the wearer more comfortable and increase freedom of movement, while the ultra-low linting material reduces environmental contamination
  • Anti-static treatment meeting EN 1149, which reduces static discharge and risk of spark and ignition
TRUSTED_SUSTAINABILITY

Specifications

Product Type Cleanroom Coveralls
Size Medium
Gender Unisex
Material Polypropylene
Color White
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Knit
Neck Style Hooded
Hood Type 3-panel
Closure Type 2-way Zipper
Seam Type Taped
Sterility Non-sterile
Resistance Bloodborne Pathogens
No. of Pockets None
For Use With (Application) Controlled Environments
Cleanroom Class Class 10-100,000 (ISO 4-8)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.