missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Protective Coverall 4565
Descrizione
- Wear 3M™ Protective Coverall 4565 for an ideal barrier to hazardous particles, limited liquid splashes and sprays to Type 4
- Fresh white coverall 4565 has recognizable red taped seams and meets a variety of standards, including chemical protective clothing EN 14126 Infective agents, EN 1073-2 Nuclear particulate resistance, EN 1149, EN ISO 13688, Type 6 Liquid Splash EN 13034 and Type 5 Dust EN ISO 13982-1, and Type 4 Liquid Spray EN 14605
- Taped seams reduce skin exposure points through tearing and reduced risk of inward leakage by chemicals or particles
- Offers Til class 2 in Particulate Radioactive Contamination EN1073-2, meaning it has very good particle hold out as well as liquid spray resistance
- Made with a non-microporous PE laminate for chemical protection with permeation-tested resistance against certain chemicals
- Knitted cuffs, elasticated waist and ankles make the wearer more comfortable and increase freedom of movement, while the ultra-low linting material reduces environmental contamination
- Anti-static treatment meeting EN 1149, which reduces static discharge and risk of spark and ignition
Specifica
Specifica
| Size | Small |
| Gender | Unisex |
| Material | Polypropylene |
| Color | White |
| Disposable | Single-use |
| Closure Type | 2-way Zipper |
| Sterility | Non-sterile |
| Resistance | Bloodborne Pathogens |
| No. of Pockets | None |
| For Use With (Application) | Controlled Environments |
| Vedi altri risultati |
Titolo del prodotto
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Individuate un'opportunità di miglioramento?