missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Protein A-HRP

Product Code. 10533205
Change view
Click to view available options
Quantity:
2 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10533205 2 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10533205 Supplier Invitrogen™ Supplier No. 101023
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Protein A-HRP

Protein A from Staphylococcus aureus, Cowan I Strain conjugated with highly purified horseradish peroxidase (RZ > 3.0).

The product is liquid containing 10 mM phosphate, 0.15 M NaCl buffer, pH 7.4, 1% ovalbumin, 40% glycerol, and 0.1% proclin.
TRUSTED_SUSTAINABILITY

Specifications

Conjugate HRP
Quantity 2 mL
Form Solution
Protein Subtype Protein A
Content And Storage Store at 2-8°C
Shipping Condition Wet Ice

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.