missing translation for 'onlineSavingsMsg'
Learn More

Novex™ Protein A - Sepharose™ 4B

Product Code. 15742456
Change view
Click to view available options
Quantity:
5 mL
25 mL
1 L
Unit Size:
1 L
25mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15742456 1 L 1 L
10727673 5 mL 5mL
10019462 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15742456 Supplier Novex™ Supplier No. 101090
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Protein A - Sepharose™ 4B

This product is prepared by covalently coupling highly purified (>98% by SDS-PAGE) Staph. aureus Cowan strain 1 Protein A to Sepharose™ 4B.

This product is supplied as a 50% suspension consisting of Protein A-Sepharose™ 4B and PBS (10 mM phosphate-buffered saline, pH 7.4, containing 0.1% Proclin).

Applications: Immunoglobulin Affinity Purification, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Product Type Protein A Sepharose
Product Line Sepharose
Column Type Affinity
Stationary Phase Protein A
Content And Storage This product is supplied as a 50% suspension (1000 ml Protein A-Sepharose™ 4B + 1000 ml PBS). Wet Ice
Quantity 1 L
Form Suspension

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.